Learn More
Teknova 2XYT Agar with 2% Glucose and Ampicillin-100. 100mm, 20 plates/sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387921
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7921 Supplier Teknova Supplier No. Y42804PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Ampicillin Sodium Salt