Learn More
Teknova 2XYT Agar Plates with 2% Glucose, Carbenicillin-100, Tetracycline-20. 245mm, 4 Plates/Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387847
Encompass
1.6% Tryptone, 1% Yeast extract, 0.5% Sodium chloride, 2% Glucose, 2% Agar, 100g/mL Carbenicillin Disodium Salt, 20g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7847 Supplier Teknova Supplier No. Y62414PK
1.6% Tryptone, 1% Yeast extract, 0.5% Sodium chloride, 2% Glucose, 2% Agar, 100g/mL Carbenicillin Disodium Salt, 20g/mL Tetracycline Hydrochloride