Learn More
Teknova 2XYT Broth. 1000mL, Sterile.
SDP
Catalog No. 502388007
Encompass
1.60% Tryptone,1.00% Yeast Extract,0.50% Sodium Chloride
Catalog No. 50-238-8007 Supplier Teknova Supplier No. Y2140
1.60% Tryptone,1.00% Yeast Extract,0.50% Sodium Chloride