Learn More
MOLTOX Molecular Toxicology Inc 2X Yeast Extract Tryptone Broth, 1000 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948773
Encompass_Preferred
This medium is a growth medium used for the cultivation of recombinant strains of E. coli. This nutritionally rich medium is also used in the propagation of M13 bacteriophage for phage display research. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-773 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26597
This medium is a growth medium used for the cultivation of recombinant strains of E. coli. This nutritionally rich medium is also used in the propagation of M13 bacteriophage for phage display research. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.