Learn More
Teknova 2XYT Broth. 1Kg. 2 pack.
SDP
Catalog No. 502387682
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride
Catalog No. 50-238-7682 Supplier Teknova Supplier No. Y02102PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride