Learn More
Teknova 2XYT Broth, 250mL. Sterile. 4 pack.
SDP
Catalog No. 502387874
Encompass
1.6% Tryptone, 1.0% Yeast Extract,0.5% Sodium Chloride
Catalog No. 50-238-7874 Supplier Teknova Supplier No. Y21674PK
1.6% Tryptone, 1.0% Yeast Extract,0.5% Sodium Chloride