Learn More
Teknova 2XYT Broth. 454g
SDP
Catalog No. 502388005
Encompass
1.6% Tryptone, 1.0% Yeast Extract,0.5% Sodium Chloride
Catalog No. 50-238-8005 Supplier Teknova Supplier No. Y0205
1.6% Tryptone, 1.0% Yeast Extract,0.5% Sodium Chloride