Learn More
Research Products International Corp 2XYT Broth, Capsules, 992 Grams of Powder in 64 Total Capsules per Bottle
SDP
Catalog No. 50489142
Encompass_Preferred
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.
2XYT Broth packaged in convenient ready to use capsules. Each capsule contains 15.5 g 2XYT Broth.
Simply add 2 capsules to one liter distilled water and heat until completely dissolved. Sterilize by autoclaving at 121°C for 15 minutes. Final pH at 25°C: 6.8-7.2.
Ingredients:
Digest Peptone: 16 g/l
Yeast Extract: 10 g/l
Sodium Chloride:5 g/l
Catalog No. 50-489-142 Supplier Research Products International Corp Supplier No. X156401000.0
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.
2XYT Broth packaged in convenient ready to use capsules. Each capsule contains 15.5 g 2XYT Broth.
Simply add 2 capsules to one liter distilled water and heat until completely dissolved. Sterilize by autoclaving at 121°C for 15 minutes. Final pH at 25°C: 6.8-7.2.
Ingredients:
Digest Peptone: 16 g/l
Yeast Extract: 10 g/l
Sodium Chloride:5 g/l