Learn More
Alkali Scientific 2XYT Broth, Liquid, 1 Liter
SDP
Catalog No. 5000660456
Encompass_Preferred
2XYT Broth, Liquid, 1 Liter, is a high-growth rate medium designed for the rapid cultivation of microorganisms. This specialized formulation provides a rich nutrient base that promotes fast replication of bacteria, making it ideal for applications requiring accelerated microbial growth. 2XYT Broth is used extensively in microbiology and molecular biology labs, especially when large numbers of bacteria are needed for experiments such as plasmid purification, gene expression, and protein production. The liquid medium supports a wide range of bacterial strains and is often used in high-density culturing, making it a go-to for laboratories focused on high-throughput bacterial growth and analysis.
Catalog No. 50-006-60456 Supplier Alkali Scientific Supplier No. XYT004CP
2XYT Broth, Liquid, 1 Liter, is a high-growth rate medium designed for the rapid cultivation of microorganisms. This specialized formulation provides a rich nutrient base that promotes fast replication of bacteria, making it ideal for applications requiring accelerated microbial growth. 2XYT Broth is used extensively in microbiology and molecular biology labs, especially when large numbers of bacteria are needed for experiments such as plasmid purification, gene expression, and protein production. The liquid medium supports a wide range of bacterial strains and is often used in high-density culturing, making it a go-to for laboratories focused on high-throughput bacterial growth and analysis.