Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2xYT Broth (Powder/Molecular Genetics), Fisher BioReagents

Catalog No. BP246710 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10,000 g
100 g
2000 g
500 g
Packaging Type:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP246710 10,000 g Poly Pail
BP2467100 100 g Poly Bottle
BP24672 2000 g Poly Bottle
BP2467500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP246710
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

For use in molecular genetics

Formulation Usage: 31.0g/L

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <4°C
Additive 2xyt Broth
Form Powder
Formulation 31g/L
For Use With (Application) Molecular Genetics
Packaging Type Poly Pail
Quantity 10,000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.