Learn More
Teknova 2XYT Broth with 0.1% glucose. 1 liter, sterile media. (Each) 2 pack.
SDP
Catalog No. 502387618
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 0.1% Cerelose Anhydrous Dextrose (Glucose)
Catalog No. 50-238-7618 Supplier Teknova Supplier No. Y01692PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 0.1% Cerelose Anhydrous Dextrose (Glucose)