Learn More
Research Products International Corp 2XYT, Powder, 500 G
SDP
Catalog No. 501978194
Encompass_Preferred
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.
Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121°C for 15 minutes. Final ph @ 25°C: 6.8-7.2.
Ingredients:
Digest Peptone: 16 g/l
Yeast Extract: 10 g/l
Sodium Chloride: 5 g/l
Catalog No. 50-197-8194 Supplier Research Products International Corp Supplier No. X15600500.0
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.
Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121°C for 15 minutes. Final ph @ 25°C: 6.8-7.2.
Ingredients:
Digest Peptone: 16 g/l
Yeast Extract: 10 g/l
Sodium Chloride: 5 g/l