Learn More
Teknova 2XYT Broth with 2.0% Glucose. 1000mL, Sterile.
SDP
Catalog No. NC0256476
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
This item is not returnable. View return policy
Catalog No. NC0256476 Supplier Teknova Supplier No. Y0162
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.