Learn More
Echelon Biosciences Research Labs 2Z 6Z-FARNESOL DIPHOSPHATE10mg
SDP
Catalog No. 50720391
Encompass
Isoprenoid compounds are a diverse group of natural products which are essential components in all cells. Isoprenoids are biosynthesized from the simple precursors isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP). Eukaryotes, fungi, and some gram-positive bacteria produce IPP through the mevalonate (MVA) pathway whereas gram-negative and some gram-positive bacteria utilize the non-mevalonate or 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway. 2Z,6Z-Farnesyl diphosphate is synthesized by Z-farnesyl diphosphate synthase (M. tuberculosis) from neryl diphosphate. It is not a substrate for protein-farnesyl transferase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-391 Supplier Echelon Biosciences Research Labs Supplier No. I017010MG
Isoprenoid compounds are a diverse group of natural products which are essential components in all cells. Isoprenoids are biosynthesized from the simple precursors isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP). Eukaryotes, fungi, and some gram-positive bacteria produce IPP through the mevalonate (MVA) pathway whereas gram-negative and some gram-positive bacteria utilize the non-mevalonate or 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway. 2Z,6Z-Farnesyl diphosphate is synthesized by Z-farnesyl diphosphate synthase (M. tuberculosis) from neryl diphosphate. It is not a substrate for protein-farnesyl transferase. This item is non returnable