Learn More
Felix Storch Inc ACCUCOLD 3.0 Cu.Ft. -85C ULT Chest Freezer
SDP
Catalog No. NC2403981
Encompass_Preferred
Manufactured for Accucold in Denmark by ELCOLD, model LAB8R6 offers 3 cu.ft. of storage. Extra thick 4 1/2' insulation and static manual defrost operation help to maintain ultra low temperatures suitable for maintaining the integrity of biological samples, industrial materials, laboratory specimens, select vaccines, and other materials with highly specific cold storage requirements. Two inner lids offer added protection by shielding contents when the outer lid is opened. The digital microprocessor controls are mounted in the front for convenient access. This unit has a temperature range that can be set between -50 and -85C, with a display viewable in Celsius or Fahrenheit. The thermostat's high/low temperature alarm warns if the interior goes out of the set range. There is also an audible 9V battery-powered alarm in the rear of the unit that will sound in the event of power failure. 6 Vertical box shelves included
Catalog No. NC2403981 Supplier Felix Storch Inc Supplier No. LAB8R6
Add to Cart
Add to Cart
Request a Quote
Manufactured for Accucold in Denmark by ELCOLD, model LAB8R6 offers 3 cu.ft. of storage. Extra thick 4 1/2' insulation and static manual defrost operation help to maintain ultra low temperatures suitable for maintaining the integrity of biological samples, industrial materials, laboratory specimens, select vaccines, and other materials with highly specific cold storage requirements. Two inner lids offer added protection by shielding contents when the outer lid is opened. The digital microprocessor controls are mounted in the front for convenient access. This unit has a temperature range that can be set between -50 and -85C, with a display viewable in Celsius or Fahrenheit. The thermostat's high/low temperature alarm warns if the interior goes out of the set range. There is also an audible 9V battery-powered alarm in the rear of the unit that will sound in the event of power failure. 6 Vertical box shelves included