Learn More
Ace Glass, Inc. Heating tape, 5/8in, 10ft length, 300w at 230v max, Glas-Col; 103A FET0.610
SDP
Catalog No. 50983298
Encompass
5/8 Glas-Col heating tape eliminates the need for a reducing voltage transformer. 10 foot length. Can be operated directly through a temperature control at normal outlet voltage. 300w at 230v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.
Catalog No. 50-983-298 Supplier Ace Glass, Inc. Supplier No. 1206312
5/8 Glas-Col heating tape eliminates the need for a reducing voltage transformer. 10 foot length. Can be operated directly through a temperature control at normal outlet voltage. 300w at 230v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.