Learn More
Aqua Solutions 3.0 mg/kg S-21 CCV Check Std. in 10:90 Oil/Xylene, 250mL
SDP
Catalog No. NC1661784
Encompass_Preferred
3.0 mg/kg S-21 CCV Check Std. in 10:90 Oil/Xylene, Boiling point/Boiling range: 36 °C (96.8 °F), Flash point: 30 °C (86 °F)
Catalog No. NC1661784 Supplier Aqua Solutions Supplier No. SH076
3.0 mg/kg S-21 CCV Check Std. in 10:90 Oil/Xylene, Boiling point/Boiling range: 36 °C (96.8 °F), Flash point: 30 °C (86 °F)