Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-(1,1-Difluoro-3-(Triisopropylsilyl)Prop-2-Yn-1-Yl)-3H-Diazirin-3-Yl)Methanol

Catalog No. 90730850MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-730-850MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 90-730-850MG Supplier Sigma Aldrich Supplier No. 90730850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-(1,1-Difluoro-3-(triisopropylsilyl)prop-2-yn-1-yl)-3H-diazirin-3-yl)methanol is a minimally interfering photo-affinity label (MI-PAL). Photoaffinity labeling is of growing interest in chemical biology and drug discovery research to characterize the non-covalent interactome and is used in protein profiling; the study of protein-protein interactions and complexes; and target engagement, identification, or validation studies. This MI-PAL contains a diazirine for UV light-induced (∼365 nm) covalent modification of a biological target with potential for downstream applications via the click chemistry-enabling alkyne tag. Its small size exhibits minimal perturbation of the interactions between proteins and the ligand or probe. The Woo lab demonstrated the electronic tuning of this MI-PAL with the placement of stability-imparting fluorine groups adjacent to the diazirine and alkyne. The alkyl fluorine ion losses are also a useful marker in collision-induced dissociation (CID) mass spectrometry experiments. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.Synthesis of an electronically-tuned minimally interfering alkynyl photo-affinity label to measure small moleculeeprotein interactions

Specifications

CAS 83152-22-1
Melting Point >300°C
Molecular Formula C17H14B2F8N4Pd
Linear Formula C17H14B2F8N4Pd
Quantity 1 g
Molecular Weight (g/mol) 554.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.