Learn More
Sklar Instruments Scissors, Bonn Iris Mini Scissors, Sklar 3-1/2 in., Sharp/Sharp, Straight, Smooth, Delicate, Stainless Steel, EA.
SDP
Catalog No. 501093945 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Bonn Iris Mini Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Delicate, Stainless Steel, EA. Sklar Bonn Miniature Iris Scissors are a finger ring ophthalmic instrument commonly used in iridectomy and other procedures involving the iris. Delicate, sharp tips, and a small profile allow the surgeon to make precise, controlled cuts. These scissors are available as both curved and straight to meet the needs of the patient and preference of the surgeon. Sklar offers these scissors with a length of 3-1/2 inches. This product is straight with smooth, sharp/sharp tips.
Catalog No. 50-109-3945 Supplier Sklar Instruments Supplier No. 642035
Scissors, Bonn Iris Mini Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Delicate, Stainless Steel, EA. Sklar Bonn Miniature Iris Scissors are a finger ring ophthalmic instrument commonly used in iridectomy and other procedures involving the iris. Delicate, sharp tips, and a small profile allow the surgeon to make precise, controlled cuts. These scissors are available as both curved and straight to meet the needs of the patient and preference of the surgeon. Sklar offers these scissors with a length of 3-1/2 inches. This product is straight with smooth, sharp/sharp tips.