Learn More
Sklar Instruments Scissors, Merit Iris Scissors, Sklar 3-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094131 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Merit Iris Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Merit Iris Scissors were originally designed for ophthalmic procedures, but have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used multiple procedures. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.
Catalog No. 50-109-4131 Supplier Sklar Instruments Supplier No. 98103
Scissors, Merit Iris Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Merit Iris Scissors were originally designed for ophthalmic procedures, but have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used multiple procedures. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.