Learn More
Sklar Instruments Scissors, Iris Scissors, Sklar 3-1/2 in., Sharp/Sharp, Straight, Smooth, Delicate, Stainless Steel, EA.
SDP
Catalog No. 501093599 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Iris Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Delicate, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. This product is straight with smooth edges, sharp tips, and a length of 3-1/2 inches.
Catalog No. 50-109-3599 Supplier Sklar Instruments Supplier No. 221135
Scissors, Iris Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Delicate, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. This product is straight with smooth edges, sharp tips, and a length of 3-1/2 inches.