Learn More
Sklar Instruments Scissors, Precision Suture Scissors, Sklar 3-1/2 in., Sharp/Sharp, Straight, Smooth, 1 Notched Blade, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 222820
Encompass_Preferred
Scissors, Precision Suture Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Straight, Smooth, 1 Notched Blade, Stainless Steel, EA. Sklar's Precision Suture Scissors are used for suture removal. This instrument has sharp tips that easily slide under and gently lift sutures in preparation for removal. One of the sharp/sharp blades has a notch that the suture rests in before it is cut. This product is straight and has an overall length of 3-1/2 inches.
Catalog No. 50-109-3648