Learn More
Sklar Instruments Scissors, Merit Spencer Stitch Scissors, Sklar 3-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094124 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Merit Spencer Stitch Scissors, Sklar, 3-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA. Merit Spencer Stitch Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2 in. and 4-1/2 in. and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.
Catalog No. 50-109-4124 Supplier Sklar Instruments Supplier No. 97346
Scissors, Merit Spencer Stitch Scissors, Sklar, 3-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA. Merit Spencer Stitch Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2 in. and 4-1/2 in. and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.