Learn More
MSC 3 1/2X4 3/8 FUEL/WATER
Catalog No. 5000502963
Encompass
Baldwin spin-on fuel filters with open ports for bowls keep fuel clean and engines running at maximum efficiency. These filters feature a port designed to be used with the OE bowl.
  • Fuel/water separator spin-on with open end for bowl
  • Contains bowl thread: 3 3/4-10 ANS Buttress; integral post seal
  • Designed for use in Racor 400, 600 Series filter assemblies
  • Compatible with Racor R45S
Catalog No. 50-005-02963 Supplier MSC Supplier No. 10957504
Baldwin spin-on fuel filters with open ports for bowls keep fuel clean and engines running at maximum efficiency. These filters feature a port designed to be used with the OE bowl.
  • Fuel/water separator spin-on with open end for bowl
  • Contains bowl thread: 3 3/4-10 ANS Buttress; integral post seal
  • Designed for use in Racor 400, 600 Series filter assemblies
  • Compatible with Racor R45S