Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one 98.0+%, TCI America™
SDP

Catalog No. B49831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49831G 1 g
B49835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49831G Supplier TCI America Supplier No. B49831G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 644-34-8
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.269
MDL Number MFCD00022990
InChI Key ATKADZVINWFQOE-SOFGYWHQSA-N
Synonym 3,4-(Methylenedioxy)chalcone
PubChem CID 5354492
IUPAC Name (E)-3-(1,3-benzodioxol-5-yl)-1-phenylprop-2-en-1-one
SMILES C1OC2=C(O1)C=C(C=C2)C=CC(=O)C3=CC=CC=C3

Specifications

Melting Point 123°C
Color Yellow
Quantity 1 g
Formula Weight 252.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one
RTECSNumber UD5572629
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.