Learn More
Sigma Aldrich 3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propanoyl chloride
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Cyclohexyl phenyl ether is an alkyl aryl ether
- It undergoes thermolysis and aquathermolysis reactions to yield 1-methylcyclopentene and phenol as major products
- Cyclohexyl phenyl ether can be synthesized from cyclohexyl bromide and phenol
- It can also be prepared from 2-cyclohexen-1-one via oxidative aromatization in the presence of vo(oet)cl2 and cyclohexanol
Specifications
Specifications
| CAS | 17137-11-0 |
| Molecular Formula | C11H8ClNO3 |
| Linear Formula | C11H8ClNO3 |
| MDL Number | MFCD00023095 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 237.64 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.