Learn More
Medchemexpress LLC 3-[1-[(3S)-1-prop-2-enoylpyrrolidin-3-yl]oxyisoquinolin-3-yl]-1,4-dihydro-1,2,4-triazol-5-one | 1627603-21-7 | 99.9% | C18H17N5O3 | 10MG
SDP
Catalog No. 5000260119
Encompass_Preferred
TAK-020 is a covalent Bruton's tyrosine kinase (BTK) inhibitor developed by Takeda and advanced as a clinical candidate for hematologic malignancies and autoimmune diseases. It irreversibly binds Cys481 in BTK's ATP-binding site and is provided by research suppliers as a characterized small-molecule reagent (CAS 1627603-21-7).
  • Covalent inhibitor that targets Cys481 in BTK.
  • Selective BTK inhibition supported by preclinical and clinical data.
  • Well-characterized small molecule with published analytical reports.
  • Available from vendors in mg-scale quantities for research use.
  • Typically supplied as high-purity material for in vitro and in vivo studies.
Catalog No. 50-002-60119 Supplier Medchemexpress LLC Supplier No. HY13287910MG
Add to Cart
Add to Cart
TAK-020 is a covalent Bruton's tyrosine kinase (BTK) inhibitor developed by Takeda and advanced as a clinical candidate for hematologic malignancies and autoimmune diseases. It irreversibly binds Cys481 in BTK's ATP-binding site and is provided by research suppliers as a characterized small-molecule reagent (CAS 1627603-21-7).
  • Covalent inhibitor that targets Cys481 in BTK.
  • Selective BTK inhibition supported by preclinical and clinical data.
  • Well-characterized small molecule with published analytical reports.
  • Available from vendors in mg-scale quantities for research use.
  • Typically supplied as high-purity material for in vitro and in vivo studies.