Learn More
Medchemexpress LLC 3-[1-(4-chlorobenzyl)-3-(t-butylthio)-5-(quinolin-2-yl-methoxy)-indol-2-yl]-2,2-dimethylpropanoic | 136668-42-3 | 99.4% | 587.17 g/mol | C34H35ClN2O3S | 50 MG
SDP
Catalog No. 5000213632
Encompass_Preferred
Quiflapon is a potent and selective inhibitor of the 5-lipoxygenase-activating protein (FLAP) used as a research tool to study leukotriene biosynthesis, inflammation, and apoptosis pathways in vitro.
  • Selective FLAP inhibitor with reported IC50 ~1.6 nM.
  • Useful for studying leukotriene biosynthesis, inflammation, and apoptosis.
  • High chemical purity (≈99.4%) suitable for biochemical assays.
  • Available as solid and as 10 mM solution in DMSO for convenience.
  • Molecular weight ~587.17 g/mol; formula C34H35ClN2O3S.
  • Supplied in multiple pack sizes to support different experimental scales.
Catalog No. 50-002-13632 Supplier Medchemexpress LLC Supplier No. HY1003750MG
Quiflapon is a potent and selective inhibitor of the 5-lipoxygenase-activating protein (FLAP) used as a research tool to study leukotriene biosynthesis, inflammation, and apoptosis pathways in vitro.
  • Selective FLAP inhibitor with reported IC50 ~1.6 nM.
  • Useful for studying leukotriene biosynthesis, inflammation, and apoptosis.
  • High chemical purity (≈99.4%) suitable for biochemical assays.
  • Available as solid and as 10 mM solution in DMSO for convenience.
  • Molecular weight ~587.17 g/mol; formula C34H35ClN2O3S.
  • Supplied in multiple pack sizes to support different experimental scales.