Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(1-(4-chlorophenyl)-4-((1H-pyrrolo[2,3-b]pyridin-4-yl)amino)piperidin-1-yl)propan-1-ol hydrochloride | 2453324-32-6 | MFCD13177916 | 99.0% | 378.30 g·mol⁻¹ | C18H21Cl2N5 | 5 MG
SDP

Catalog No. 5000262961
Encompass_Preferred

CCT128930 hydrochloride is the hydrochloride salt of a potent and selective AKT (protein kinase B) inhibitor for preclinical research. Supplied as a solid, it is intended for biochemical and cellular studies where high potency and defined physicochemical properties are required.

  • Potent and selective AKT inhibitor (AKT IC50 ≈ 6 nM).
  • Hydrochloride salt; solid, white to yellow.
  • High purity (≈99.0%).
  • Molecular formula C18H21Cl2N5; molecular weight 378.30 g·mol⁻¹.
  • Soluble in DMSO up to 20 mg/mL with warming and sonication.
  • Recommended storage sealed, away from moisture; in solution -80°C (≤6 months).
  • Supplied in small research quantities suitable for assay development.

Catalog No. 50-002-62961 Supplier Medchemexpress LLC Supplier No. HY13260A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

CCT128930 hydrochloride is the hydrochloride salt of a potent and selective AKT (protein kinase B) inhibitor for preclinical research. Supplied as a solid, it is intended for biochemical and cellular studies where high potency and defined physicochemical properties are required.

  • Potent and selective AKT inhibitor (AKT IC50 ≈ 6 nM).
  • Hydrochloride salt; solid, white to yellow.
  • High purity (≈99.0%).
  • Molecular formula C18H21Cl2N5; molecular weight 378.30 g·mol⁻¹.
  • Soluble in DMSO up to 20 mg/mL with warming and sonication.
  • Recommended storage sealed, away from moisture; in solution -80°C (≤6 months).
  • Supplied in small research quantities suitable for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.