Learn More
3-(1,5-Dibenzyl-1H-indazole-3-yl)oxypropyldimethylamine hydrochloride, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
- This product is provided as delivered and specified by the issuing Pharmacopoeia
- All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 2196183-71-6 |
| Flash Point | Not Applicable |
| Molecular Formula | C26H29N3O · HCl |
| Linear Formula | C26H29N3O · HCl |
| Quantity | 25 mg |
| Formula Weight | 435.99 |
| Grade | BP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.