Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(1-Aminoethyl)phenol | 63720-38-7 | 98.0% | C8H11NO | 5 G
SDP

Catalog No. 5000434764
Encompass_Preferred

3-(1-Aminoethyl)phenol is a chemical compound with the molecular formula C8H11NO and a molecular weight of 137.18. It appears as a white to off-white solid with a purity of 97.98% by LCMS analysis. This product is suitable for various laboratory applications.

  • White to off-white solid
  • High purity (97.98%)
  • Consistent with structure by 1H NMR and LCMS
  • Long-term stability when stored as powder at -20°C or 4°C
  • Stable in solvent when stored at -80°C or -20°C

Catalog No. 50-004-34764 Supplier Medchemexpress LLC Supplier No. HY700315G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(1-Aminoethyl)phenol is a chemical compound with the molecular formula C8H11NO and a molecular weight of 137.18. It appears as a white to off-white solid with a purity of 97.98% by LCMS analysis. This product is suitable for various laboratory applications.

  • White to off-white solid
  • High purity (97.98%)
  • Consistent with structure by 1H NMR and LCMS
  • Long-term stability when stored as powder at -20°C or 4°C
  • Stable in solvent when stored at -80°C or -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.