Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-[(1-Benzyl-4-piperidinyl)amino]-1-propanol

Catalog No. CDS002476 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002476 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002476 Supplier Sigma Aldrich Supplier No. CDS00247650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Benzyl sulfone is a cyclic sulfone
  • Benzyl sulfone (dibenzyl sulfone) on pyrolysis at 290c yields stillbene and toluene
  • Mechanism of pyrolysis of dibenzyl sulfones at 600-700c has been reported
  • Benzyl sulfone forms sodium salt with metallic sodium and with sodium ethoxide
  • Cryoscopic behavior of benzyl sulfone was investigated

Specifications

Molecular Formula C15H24N2O
Linear Formula C15H24N2O
MDL Number MFCD05237180
Quantity 10 g
Molecular Weight (g/mol) 248.36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.