missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 3-(1-Ethyl-1H-pyrazol-4-yl)propan-1-ol

Catalog No. CBR003791G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBR003791G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBR003791G Supplier Sigma Aldrich Supplier No. CBR003791G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(1-Ethyl-1H-pyrazol-4-yl)propan-1-ol

Specifications

CAS 1007516-30-4
Molecular Formula C8H14N2O
Linear Formula C8H14N2O
MDL Number MFCD08059737
Quantity 250 mg
Molecular Weight (g/mol) 154.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.