Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC60603DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC60603DA 1 g Amber Glass Bottle
CC60603CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC60603DA Supplier Thermo Scientific Chemicals Supplier No. CC60603DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 912569-59-6
Molecular Formula C10H9ClN2O2S
Molecular Weight (g/mol) 256.704
MDL Number MFCD09817503
InChI Key QGUULJKRHYTFTP-UHFFFAOYSA-N
PubChem CID 24229628
IUPAC Name 3-(1-methylpyrazol-3-yl)benzenesulfonyl chloride
SMILES CN1C=CC(=N1)C2=CC(=CC=C2)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-1-methyl-1h-pyrazol-3-yl benzenesulfonyl chloride, benzenesulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl benzenesulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl benzenesulphonyl chloride, 3-1-methyl-1h-pyrazol-3-yl benzene-1-sulfonyl chloride, chloro 3-1-methylpyrazol-3-yl phenyl sulfone, 3-1-methyl-1h-pyrazol-3-yl phenylsulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl-benzenesulfonyl chloride, benzenesulfonyl chloride,3-1-methyl-1h-pyrazol-3-yl
Formula Weight 256.71
Percent Purity 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.