Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(1-Naphthoyl)indole | 109555-87-5 | 96.67% | C19H13NO | 1 G
SDP

Catalog No. 5000433639
Encompass_Preferred

3-(1-Naphthoyl)indole is a biological molecule primarily intended for research use. It presents as a white to pink solid with a purity of 96.67%.

  • Solid appearance, white to pink color
  • High purity of 96.67%
  • Molecular weight of 271.31
  • Melting point at 236°C
  • Boiling point at 512.2°C
  • Stable for extended periods when stored as powder or in solvent under recommended conditions
  • Suitable for laboratory research applications

Catalog No. 50-004-33639 Supplier Medchemexpress LLC Supplier No. HY750041G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(1-Naphthoyl)indole is a biological molecule primarily intended for research use. It presents as a white to pink solid with a purity of 96.67%.

  • Solid appearance, white to pink color
  • High purity of 96.67%
  • Molecular weight of 271.31
  • Melting point at 236°C
  • Boiling point at 512.2°C
  • Stable for extended periods when stored as powder or in solvent under recommended conditions
  • Suitable for laboratory research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.