Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(1-Naphthoyl)indole | 109555-87-5 | MFCD12923093 | 96.67% | C19H13NO | 250 MG
SDP

Catalog No. 5000358439
Encompass_Preferred

3-(1-Naphthoyl)indole is a chemical compound supplied by MedChemExpress, primarily used as a biochemical assay reagent. This biological molecule has a molecular formula of C19H13NO and a molecular weight of 271.31. It is identified by its CAS number 109555-87-5 and MFCD number MFCD12923093. This high-purity compound, typically appearing as a white to pink solid, requires specific storage conditions to maintain its integrity, ensuring its stability for various research applications.

  • Purity of 96.67%
  • Used as a biochemical assay reagent
  • Molecular weight of 271.31
  • Appears as a white to pink solid
  • Recommended storage for powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month
  • Shipped at room temperature if less than 2 weeks

Catalog No. 50-003-58439 Supplier Medchemexpress LLC Supplier No. 250MGHY75004
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(1-Naphthoyl)indole is a chemical compound supplied by MedChemExpress, primarily used as a biochemical assay reagent. This biological molecule has a molecular formula of C19H13NO and a molecular weight of 271.31. It is identified by its CAS number 109555-87-5 and MFCD number MFCD12923093. This high-purity compound, typically appearing as a white to pink solid, requires specific storage conditions to maintain its integrity, ensuring its stability for various research applications.

  • Purity of 96.67%
  • Used as a biochemical assay reagent
  • Molecular weight of 271.31
  • Appears as a white to pink solid
  • Recommended storage for powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month
  • Shipped at room temperature if less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.