Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1-Naphthyl)-D-alanine, 95%, Thermo Scientific™

Catalog No. AAH6640803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6640803 1 g
AAH6640806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6640803 Supplier Thermo Scientific Chemicals Supplier No. H6640803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(1-Naphthyl)-D-alanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in hydrochloric acid.

Notes
Protect from heat. Store away from oxidizing agents. Refrigerate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78306-92-0
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD00038544
InChI Key OFYAYGJCPXRNBL-GFCCVEGCSA-N
Synonym d-1-naphthylalanine, 3-1-naphthyl-d-alanine, h-d-1-nal-oh, r-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-d-ala, 2r-2-amino-3-naphthalen-1-ylpropanoic acid, d-3-1-naphthyl alanine, 2r-2-amino-3-naphthalen-1-yl propanoic acid, 2r-2-amino-3-naphth-1-yl propanoic acid, 3-naphth-1-yl-d-alanine
PubChem CID 6950505
IUPAC Name (2R)-2-amino-3-naphthalen-1-ylpropanoic acid
SMILES C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)N

Specifications

Beilstein 4352834
Solubility Information Soluble in hydrochloric acid.
Quantity 1 g
Formula Weight 215.25
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(1-Naphthyl)-D-alanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.