Learn More
Polysciences, Inc. Glass Beads (3 to 10um), From Specially Selected Soda Lime Glass, CAS No.: 65997-17-3, Store at 25 deg.C, Specific Gravity: 2.5, Light Green Odorless Beads, 1g
SDP
Catalog No. NC9154972
Encompass
Glass Beads (3 to 10um), From Specially Selected Soda Lime Glass, CAS No.: 65997-17-3, Store at 25 deg.C, Specific Gravity: 2.5, Light Green Odorless Beads, 1g
Catalog No. NC9154972 Supplier Polysciences, Inc. Supplier No. 076661
Glass Beads (3 to 10um), From Specially Selected Soda Lime Glass, CAS No.: 65997-17-3, Store at 25 deg.C, Specific Gravity: 2.5, Light Green Odorless Beads, 1g