Learn More
Medchemexpress LLC 3-(10′-Phenothiazinyl)propane-1-sulfonate sodium | 101199-38-6 | 99.6% | 343.40 | 50 MG
SDP
Catalog No. 5000435614
Encompass_Preferred
PTZ-343 is a potent enhancer of Luminol. It significantly increases the light output of the peroxidase-catalyzed luminol chemiluminescent oxidation reaction (>80%). PTZ-343 acts as an electron transfer mediator, reacting with HRP-II to release HRP and an enhancer (PTZ-343) radical, which then rapidly oxidizes luminol anions to induce light emission. For research use only.
  • Potent enhancer of luminol
  • Greatly increases light output of peroxidase-catalyzed luminol chemiluminescent oxidation reaction
  • Acts as an electron transfer mediator
  • Induces light emission through rapid oxidation of luminol anions
Catalog No. 50-004-35614 Supplier Medchemexpress LLC Supplier No. HY11284150MG
PTZ-343 is a potent enhancer of Luminol. It significantly increases the light output of the peroxidase-catalyzed luminol chemiluminescent oxidation reaction (>80%). PTZ-343 acts as an electron transfer mediator, reacting with HRP-II to release HRP and an enhancer (PTZ-343) radical, which then rapidly oxidizes luminol anions to induce light emission. For research use only.
  • Potent enhancer of luminol
  • Greatly increases light output of peroxidase-catalyzed luminol chemiluminescent oxidation reaction
  • Acts as an electron transfer mediator
  • Induces light emission through rapid oxidation of luminol anions