Learn More
Ace Glass, Inc. Combo mantle power control, 3 position, 120v 15amp output, percentage timer (pulse) type
SDP
Catalog No. 50983290
Encompass
3-position Power Control for multi-element combination heating mantle. Power control is the percentage timer (pulse) type. 120v 15amp output.
Catalog No. 50-983-290 Supplier Ace Glass, Inc. Supplier No. 1206161
3-position Power Control for multi-element combination heating mantle. Power control is the percentage timer (pulse) type. 120v 15amp output.