Learn More
Ace Glass, Inc. Combo mantle power control, 3 position, 120v 15amp output, proportional voltage (steady state) type
SDP
Catalog No. 50983291
Encompass
3-position Power Control for multi-element combination heating mantle. Power control is the proportional voltage (steady state) type. 120v 15amp output.
Catalog No. 50-983-291 Supplier Ace Glass, Inc. Supplier No. 1206171
3-position Power Control for multi-element combination heating mantle. Power control is the proportional voltage (steady state) type. 120v 15amp output.