Learn More
Medchemexpress LLC 3-(15-hydroxypentadecyl)-2,4,4-trimethylcyclohex-2-en-1-one | 220757-88-0 | MFCD09832597 | 99.1% | 364.60 g/mol | C24H44O2 | 100 MG
SDP
Catalog No. 5000242416
Encompass_Preferred
tCFA15 is a trimethyl cyclohexenonic long-chain fatty alcohol with a 15-carbon side chain. It is reported to promote neuronal differentiation and may regulate Notch signaling. Supplied as a research/analytical standard, the material is accompanied by batch-specific quality documentation and is intended for laboratory research use only.
  • Trimethyl cyclohexenonic long-chain fatty alcohol structure.
  • Contains a 15-carbon side chain.
  • Promotes neuronal differentiation in reported studies.
  • May modulate the Notch signaling pathway.
  • High purity suitable for analytical and research applications.
  • Provided with batch-specific certificate of analysis.
Catalog No. 50-002-42416 Supplier Medchemexpress LLC Supplier No. HY104031100MG
tCFA15 is a trimethyl cyclohexenonic long-chain fatty alcohol with a 15-carbon side chain. It is reported to promote neuronal differentiation and may regulate Notch signaling. Supplied as a research/analytical standard, the material is accompanied by batch-specific quality documentation and is intended for laboratory research use only.
  • Trimethyl cyclohexenonic long-chain fatty alcohol structure.
  • Contains a 15-carbon side chain.
  • Promotes neuronal differentiation in reported studies.
  • May modulate the Notch signaling pathway.
  • High purity suitable for analytical and research applications.
  • Provided with batch-specific certificate of analysis.