Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(15-hydroxypentadecyl)-2,4,4-trimethylcyclohex-2-en-1-one | 220757-88-0 | MFCD09832597 | 99.1% | 364.60 g/mol | C24H44O2 | 50 MG
SDP

Catalog No. 5000243938
Encompass_Preferred

tCFA15 is a trimethyl cyclohexenonic long-chain fatty alcohol (IUPAC: 3-(15-hydroxypentadecyl)-2,4,4-trimethylcyclohex-2-en-1-one; CAS 220757-88-0) used as a research reagent to promote neuronal differentiation and modulate Notch signaling. It is supplied as an analytical standard and characterized for in vitro biochemical and cell-based assays.

  • High purity (99.12% by LCMS).
  • Molecular weight 364.60 g/mol.
  • Soluble in DMSO.
  • Suitable for neuronal differentiation and Notch signaling studies.
  • Provided as an analytical standard for HPLC, GC, and MS methods.

Catalog No. 50-002-43938 Supplier Medchemexpress LLC Supplier No. HY10403150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tCFA15 is a trimethyl cyclohexenonic long-chain fatty alcohol (IUPAC: 3-(15-hydroxypentadecyl)-2,4,4-trimethylcyclohex-2-en-1-one; CAS 220757-88-0) used as a research reagent to promote neuronal differentiation and modulate Notch signaling. It is supplied as an analytical standard and characterized for in vitro biochemical and cell-based assays.

  • High purity (99.12% by LCMS).
  • Molecular weight 364.60 g/mol.
  • Soluble in DMSO.
  • Suitable for neuronal differentiation and Notch signaling studies.
  • Provided as an analytical standard for HPLC, GC, and MS methods.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.