Learn More
Electronic Imaging Materials Inc 3.15 Inches x 300 Meter T68 Premium Resin Ribbon - EIMINC
SDP
Catalog No. NC1160127
Encompass_Preferred
T68—3.15 inches x 300 meter resin ribbon specially formulated to print exceptional images. Handles some of the harshest environmental conditions—including resistance against abrasion, heat, steam and chemicals such as gasoline, mineral spirits, oil, bleach, IPA, antifreeze, and sulfuric acid.*Note: make sure to find out if ribbon is compatible with your printer & label material before purchasing. Works in Zebra printer models ZT220/230, ZT410/420, ZD420, as well as TSC TTP247/345 and TSC TX series printers.
Catalog No. NC1160127 Supplier Electronic Imaging Materials Inc Supplier No. T68315300
T68—3.15 inches x 300 meter resin ribbon specially formulated to print exceptional images. Handles some of the harshest environmental conditions—including resistance against abrasion, heat, steam and chemicals such as gasoline, mineral spirits, oil, bleach, IPA, antifreeze, and sulfuric acid.*Note: make sure to find out if ribbon is compatible with your printer & label material before purchasing. Works in Zebra printer models ZT220/230, ZT410/420, ZD420, as well as TSC TTP247/345 and TSC TX series printers.