Learn More
BRANSON ULTRASONICS CORPORATION (3P) 3/16IN MICROTIP SLP UNIT ONLY
Catalog No. NC3721536
Encompass_Preferred
Branson Ultrasonics Microtips for Sonifiers SLPe, SLPt, 150E and 150T, Diameter: 0.19 in., 0.48cm, Capacity: 5 to 25mL, Volume: 5 to 25mL, For Use With: SLPe, SLPt, 150E and 150T Sonifier, Ideal for processing samples to the smallest diameter vessels, SLP Microtip, Length: 4 in., 10.16cm
Catalog No. NC3721536 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 1091221182
Branson Ultrasonics Microtips for Sonifiers SLPe, SLPt, 150E and 150T, Diameter: 0.19 in., 0.48cm, Capacity: 5 to 25mL, Volume: 5 to 25mL, For Use With: SLPe, SLPt, 150E and 150T Sonifier, Ideal for processing samples to the smallest diameter vessels, SLP Microtip, Length: 4 in., 10.16cm