Learn More
Medchemexpress LLC (3β)-17-{[(Trifluoromethyl)sulfonyl]oxy}androsta-5,16-dien-3-yl acetate | 115375-60-5 | 99.09% | C22H29F3O5S | 1 G
SDP
Catalog No. 5000434324
Encompass_Preferred
(3S,8R,9S,10R,13S,14S)-10,13-Dimethyl-17-(trifluoromethylsulfonyloxy)-2,3,4,7,8,9,10,11,12,13,14,15-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl acetate, also known as ((3β)-17-{[(Trifluoromethyl)sulfonyl]oxy}androsta-5,16-dien-3-yl acetate), is a drug intermediate. It is used for the synthesis of various active compounds.
  • Drug intermediate
  • Used for synthesis of various active compounds
  • Target: drug intermediate
  • Pathway: others
Catalog No. 50-004-34324 Supplier Medchemexpress LLC Supplier No. HY756241G
(3S,8R,9S,10R,13S,14S)-10,13-Dimethyl-17-(trifluoromethylsulfonyloxy)-2,3,4,7,8,9,10,11,12,13,14,15-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl acetate, also known as ((3β)-17-{[(Trifluoromethyl)sulfonyl]oxy}androsta-5,16-dien-3-yl acetate), is a drug intermediate. It is used for the synthesis of various active compounds.
  • Drug intermediate
  • Used for synthesis of various active compounds
  • Target: drug intermediate
  • Pathway: others