Learn More
Medchemexpress LLC (3β)-17-{[(Trifluoromethyl)sulfonyl]oxy}androsta-5,16-dien-3-yl acetate | 115375-60-5 | 99.1% | C22H29F3O5S | 500 MG
SDP
Catalog No. 5000433755
Encompass_Preferred
This chemical is a drug intermediate used for the synthesis of various active compounds.
  • High purity (99.1%)
  • Molecular weight of 462.52
  • Appears as a solid
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Intended for research use only
Catalog No. 50-004-33755 Supplier Medchemexpress LLC Supplier No. HY75624500MG
This chemical is a drug intermediate used for the synthesis of various active compounds.
  • High purity (99.1%)
  • Molecular weight of 462.52
  • Appears as a solid
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Intended for research use only