Learn More
Medchemexpress LLC 3-(1H-benzimidazol-1-yl)propan-1-amine | 73866-15-6 | MFCD03093044 | 99.9% | 175.24 g·mol⁻¹ | C10H13N3 | 100 MG
SDP
Catalog No. 5000299821
Encompass_Preferred
3-(1H-Benzimidazol-1-yl)propan-1-amine is a benzimidazole-containing propylamine used primarily as a chemical intermediate in medicinal chemistry and organic synthesis. Identified by CAS 73866-15-6, it has formula C10H13N3 and molecular weight 175.24 g·mol⁻¹. The material is supplied for research use only and is typically handled as a powder under refrigerated storage to preserve integrity.
  • Benzimidazole scaffold suitable for medicinal chemistry synthesis.
  • Useful as a building block for heterocyclic compound libraries.
  • Available in small-scale lab pack sizes for route scouting.
  • High reported purity supporting analytical and synthetic work.
  • Molecular weight 175.24 g·mol⁻¹ and formula C10H13N3.
  • Stored as a powder at 4°C, protect from light.
  • Supplied for research use only (not for human use).
Catalog No. 50-002-99821 Supplier Medchemexpress LLC Supplier No. HYW039840100MG
Add to Cart
Add to Cart
3-(1H-Benzimidazol-1-yl)propan-1-amine is a benzimidazole-containing propylamine used primarily as a chemical intermediate in medicinal chemistry and organic synthesis. Identified by CAS 73866-15-6, it has formula C10H13N3 and molecular weight 175.24 g·mol⁻¹. The material is supplied for research use only and is typically handled as a powder under refrigerated storage to preserve integrity.
  • Benzimidazole scaffold suitable for medicinal chemistry synthesis.
  • Useful as a building block for heterocyclic compound libraries.
  • Available in small-scale lab pack sizes for route scouting.
  • High reported purity supporting analytical and synthetic work.
  • Molecular weight 175.24 g·mol⁻¹ and formula C10H13N3.
  • Stored as a powder at 4°C, protect from light.
  • Supplied for research use only (not for human use).