Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(1H-benzimidazol-1-yl)propan-1-amine | 73866-15-6 | MFCD03093044 | 99.9% | 175.24 g·mol⁻¹ | C10H13N3 | 5 G
SDP

Catalog No. 5000301963
Encompass_Preferred

3-(1H-Benzimidazol-1-yl)propan-1-amine is a benzimidazole-containing small-molecule intermediate used in the synthesis of pharmaceutical and research compounds. Supplied for research use, it is characterized by high purity and should be stored and handled under controlled conditions to maintain stability and integrity.

  • Used as a drug intermediate for chemical synthesis
  • Molecular formula C10H13N3
  • Molecular weight 175.24 g·mol⁻¹
  • Physical appearance: liquid
  • High purity (99.89% by LCMS)
  • Recommended storage 4°C; protect from light; in solvent: -80°C (6 months), -20°C (1 month)
  • Intended for research use only

Catalog No. 50-003-01963 Supplier Medchemexpress LLC Supplier No. HYW0398405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(1H-Benzimidazol-1-yl)propan-1-amine is a benzimidazole-containing small-molecule intermediate used in the synthesis of pharmaceutical and research compounds. Supplied for research use, it is characterized by high purity and should be stored and handled under controlled conditions to maintain stability and integrity.

  • Used as a drug intermediate for chemical synthesis
  • Molecular formula C10H13N3
  • Molecular weight 175.24 g·mol⁻¹
  • Physical appearance: liquid
  • High purity (99.89% by LCMS)
  • Recommended storage 4°C; protect from light; in solvent: -80°C (6 months), -20°C (1 month)
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.