Learn More
Medchemexpress LLC 3-(1H-benzimidazol-2-yl)-7-(diethylamino)chromen-2-one | 27425-55-4 | MFCD00051348 | 99.8% | 333.38 g/mol | C20H19N3O2 | 100MG
SDP
Catalog No. 5000234918
Encompass_Preferred
Coumarin 7 (CAS 27425-55-4) is a coumarin-class laser dye and fluorescent research reagent used in spectroscopy and laser applications. It has molecular formula C20H19N3O2, a molecular weight of 333.38 g/mol, and is supplied with a reported purity of 99.80% for laboratory use.
  • High reported purity: 99.8%.
  • Suitable for spectroscopy, fluorescence, and laser dye studies.
  • Molecular formula C20H19N3O2 and molecular weight 333.38 g/mol.
  • Available in small research quantities, including 50 mg and 100 mg.
  • Intended for research use in laboratory applications.
Catalog No. 50-002-34918 Supplier Medchemexpress LLC Supplier No. HY125750100MG
Coumarin 7 (CAS 27425-55-4) is a coumarin-class laser dye and fluorescent research reagent used in spectroscopy and laser applications. It has molecular formula C20H19N3O2, a molecular weight of 333.38 g/mol, and is supplied with a reported purity of 99.80% for laboratory use.
  • High reported purity: 99.8%.
  • Suitable for spectroscopy, fluorescence, and laser dye studies.
  • Molecular formula C20H19N3O2 and molecular weight 333.38 g/mol.
  • Available in small research quantities, including 50 mg and 100 mg.
  • Intended for research use in laboratory applications.